siRNA Details
Gene Symbol: NAA20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016100.4 | NP_057184.1 | 4, 5 | 406 | ||
NM_181527.3 | NP_852668.1 | 4, 5 | 493 | |||
GenBank mRNA | AF085355.1 | - | 4, 5 | 369 | ||
AK311819.1 | - | 4, 5 | 359 | |||
AL136641.1 | - | 4, 5 | 338 | |||
BC005181.1 | - | 4, 5 | 334 | 3950192 | ||
BC008446.1 | - | 4, 5 | 355 | 4104421 | ||
CB959658.1 | - | 4, 5 | 430 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
