siRNA Details
Gene Symbol: DSCR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026838.1 | 4 | 685 | |||
NR_026839.1 | 3 | 588 | ||||
NR_026840.1 | 4 | 446 | ||||
NR_026841.1 | 4 | 428 | ||||
NR_026842.1 | 3 | 331 | ||||
GenBank mRNA | AF321193.1 | - | 4 | 403 | ||
AF426256.1 | - | 4 | 351 | |||
AJ001904.1 | - | 4 | 388 | |||
AJ297914.2 | - | 4 | 406 | |||
AJ301615.2 | - | 3 | 309 | |||
AJ305085.2 | - | 3 | 566 | |||
AJ306839.2 | - | 4 | 424 | |||
AJ306840.2 | - | 4 | 478 | |||
AJ783421.1 | - | 4 | 663 | |||
BC015981.1 | - | 3 | 313 | |||
BC029353.1 | - | 3 | 588 | |||
BC070229.1 | - | 3 | 290 | |||
BC117304.1 | - | 4 | 360 | |||
BC117330.1 | - | 4 | 360 | |||
BC143995.1 | - | 4 | 378 | |||
BG189423.1 | - | 1 | 83 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
