siRNA Details
Gene Symbol: DSCR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026838.1 | 2 | 231 | |||
NR_026839.1 | 2 | 231 | ||||
NR_026840.1 | 2 | 231 | ||||
NR_026841.1 | 2 | 231 | ||||
NR_026842.1 | 2 | 231 | ||||
GenBank mRNA | AF321193.1 | - | 2 | 206 | ||
AF426256.1 | - | 2 | 154 | |||
AJ001904.1 | - | 2 | 191 | |||
AJ297914.2 | - | 2 | 209 | |||
AJ301615.2 | - | 2 | 209 | |||
AJ305085.2 | - | 2 | 209 | |||
AJ306839.2 | - | 2 | 209 | |||
AJ306840.2 | - | 2 | 209 | |||
AJ783421.1 | - | 2 | 209 | |||
BC015981.1 | - | 2 | 213 | |||
BC029353.1 | - | 2 | 231 | |||
BC070229.1 | - | 2 | 190 | |||
BC117304.1 | - | 2 | 163 | |||
BC117330.1 | - | 2 | 163 | |||
BC143995.1 | - | 2 | 163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
