siRNA Details
Gene Symbol: RTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321859.1 | NP_001308788.1 | 5, 6 | 2899 | ||
NM_001321860.1 | NP_001308789.1 | 5, 6 | 2969 | |||
NM_001321861.1 | NP_001308790.1 | 5, 6 | 2932 | |||
NM_001321862.1 | NP_001308791.1 | 5, 6 | 2919 | |||
NM_001321863.1 | NP_001308792.1 | 5, 6 | 2869 | |||
NM_001321904.1 | NP_001308833.1 | 6, 7 | 3347 | |||
NM_007008.2 | NP_008939.1 | 3, 4 | 589 | |||
NM_020532.4 | NP_065393.1 | 5, 6 | 3652 | |||
NM_153828.2 | NP_722550.1 | 3, 4 | 1195 | |||
NM_207520.1 | NP_997403.1 | 4, 5 | 1252 | |||
NM_207521.1 | NP_997404.1 | 5, 6 | 2910 | |||
NR_135829.1 | 3, 4 | 475 | ||||
NR_135830.1 | 3, 4 | 453 | ||||
XM_005264434.3 | XP_005264491.1 | 6, 7 | 3046 | |||
XM_017004518.1 | XP_016860007.1 | 6, 7 | 3085 | |||
XM_017004519.1 | XP_016860008.1 | 6, 7 | 3202 | |||
GenBank mRNA | AB015639.1 | - | 3, 4 | 963 | ||
AB020693.1 | - | 5, 6 | 3488 | |||
AB040462.1 | - | 5, 6 | 3598 | |||
AB040463.1 | - | 3, 4 | 1141 | |||
AF063601.1 | - | 3, 4 | 1912 | |||
AF077050.1 | - | 3, 4 | 588 | |||
AF087901.1 | - | 3, 4 | 480 | |||
AF125103.1 | - | 3, 4 | 589 | |||
AF132047.1 | - | 3, 4 | 1076 | |||
AF132048.1 | - | 3, 4 | 495 | |||
AF148537.1 | - | 5, 6 | 3495 | |||
AF148538.1 | - | 3, 4 | 1038 | |||
AF177332.1 | - | 3, 4 | 494 | |||
AF320999.1 | - | 4, 5 | 2658 | |||
AF333336.1 | - | 5, 6 | 2914 | |||
AJ251383.1 | - | 5, 6 | 3354 | |||
AJ251384.1 | - | 3, 4 | 897 | |||
AJ251385.1 | - | 3, 4 | 375 | |||
AK075039.1 | - | 3, 4 | 1129 | |||
AK129806.1 | - | 3, 4 | 1054 | |||
AK130812.1 | - | 3, 4 | 956 | |||
AK172726.1 | - | 1, 2 | 19 | |||
AY102276.1 | - | 3, 4 | 609 | |||
AY102277.1 | - | 3, 4 | 1141 | |||
AY102278.1 | - | 4, 5 | 1198 | |||
AY102279.1 | - | 5, 6 | 3598 | |||
AY123245.1 | - | 5, 6 | 2911 | |||
AY123246.1 | - | 5, 6 | 2969 | |||
AY123247.1 | - | 5, 6 | 2932 | |||
AY123248.1 | - | 5, 6 | 2918 | |||
AY123249.1 | - | 5, 6 | 2879 | |||
AY123250.1 | - | 5, 6 | 2869 | |||
BC001035.2 | - | 3, 4 | 560 | |||
BC007109.1 | - | 3, 4 | 486 | |||
BC010737.1 | - | 3, 4 | 902 | |||
BC012619.1 | - | 3, 4 | 1052 | |||
BC014366.1 | - | 3, 4 | 486 | |||
BC016165.1 | - | 4, 5 | 1201 | |||
BC026788.1 | - | 3, 4 | 1083 | |||
BC068991.1 | - | 3, 4 | 1145 | |||
BC071848.1 | - | 4, 5 | 883 | |||
BC150182.1 | - | 5, 6 | 3602 | 40148210 | ||
BC152425.1 | - | 5, 6 | 3488 | 8862539 | ||
BC152555.1 | - | 5, 6 | 3602 | 40148984 | ||
CN429712.1 | - | 3, 4 | 424 | |||
DQ778739.1 | - | 3, 4 | 375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
