siRNA Details
Gene Symbol: CEACAM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002483.6 | NP_002474.4 | 6 | 2543 | ||
XM_011526990.1 | XP_011525292.1 | 5 | 2525 | |||
GenBank mRNA | M18216.1 | - | 6 | 2469 | ||
M18728.1 | - | 6 | 2475 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
