siRNA Details
Gene Symbol: GOLGA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002296.1 | NP_001002296.1 | 2 | 429 | ||
NM_001174124.1 | NP_001167595.1 | 3 | 306 | |||
NM_016099.2 | NP_057183.2 | 3 | 306 | |||
GenBank mRNA | AB104615.1 | - | 3 | 286 | ||
AF068291.1 | - | 2 | 260 | |||
AF125102.1 | - | 3 | 270 | |||
AK026613.1 | - | 3 | 286 | |||
AL695304.1 | - | 3 | 315 | |||
BC012032.1 | - | 2 | 429 | 4707385 | ||
BI553491.1 | - | 3 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
