siRNA Details
Gene Symbol: GOLGA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002296.1 | NP_001002296.1 | 5 | 1724 | ||
NM_001174124.1 | NP_001167595.1 | 6 | 1606 | |||
NM_016099.2 | NP_057183.2 | 6 | 1601 | |||
GenBank mRNA | AB104615.1 | - | 6 | 1581 | ||
AF125102.1 | - | 6 | 1565 | |||
AK026613.1 | - | 6 | 1586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
