siRNA Details
Gene Symbol: TRAPPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011658.3 | NP_001011658.1 | 6 | 2237 | ||
NM_001128835.2 | NP_001122307.2 | 6 | 2231 | |||
NM_014563.5 | NP_055378.1 | 5 | 2095 | |||
XM_011545565.1 | XP_011543867.1 | 7 | 2305 | |||
XM_011545566.2 | XP_011543868.1 | 6 | 2161 | |||
GenBank mRNA | AK055671.1 | - | 1 | 1402 | ||
BC016915.1 | - | 6 | 1361 | 4432196 | ||
BC052618.1 | - | 5 | 2068 | 6581382 | ||
BX112362.1 | - | 1 | 253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
