siRNA Details
Gene Symbol: SCFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283031.1 | NP_001269960.1 | 2, 3 | 225 | ||
NM_001283033.1 | NP_001269962.1 | 2, 3 | 225 | |||
NM_016106.3 | NP_057190.2 | 2, 3 | 225 | |||
XM_005267469.2 | XP_005267526.1 | 2, 3 | 223 | |||
XM_011536586.2 | XP_011534888.1 | 2, 3 | 222 | |||
XM_011536587.2 | XP_011534889.1 | 2, 3 | 222 | |||
XM_011536588.2 | XP_011534890.1 | 2, 3 | 222 | |||
XM_011536589.1 | XP_011534891.1 | 2, 3 | 222 | |||
XM_017021113.1 | XP_016876602.1 | 2, 3 | 222 | |||
GenBank mRNA | AB020724.2 | - | 2, 3 | 160 | ||
AF086916.1 | - | 2, 3 | 148 | |||
AF092139.1 | - | 2, 3 | 141 | |||
AF110646.1 | - | 2, 3 | 136 | |||
AF319958.1 | - | 2, 3 | 164 | |||
AK222820.1 | - | 2, 3 | 144 | |||
AK290410.1 | - | 2, 3 | 160 | |||
AK299222.1 | - | 2, 3 | 152 | |||
AK307960.1 | - | 2, 3 | 160 | |||
AK308587.1 | - | 2, 3 | 168 | |||
BC017734.1 | - | 2, 3 | 140 | 4420306 | ||
BU198450.1 | - | 2, 3 | 166 | |||
CA454055.1 | - | 2, 3 | 151 | |||
CD697934.1 | - | 2, 3 | 189 | |||
DA151789.1 | - | 2, 3 | 223 | |||
DN993082.1 | - | 2, 3 | 147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
