siRNA Details
Gene Symbol: HIST1H2BB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021062.2 | NP_066406.1 | 1 | 17 | ||
GenBank mRNA | BC096728.1 | - | 1 | 17 | 40014250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
