siRNA Details
Gene Symbol: PCDH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318372.1 | NP_001305301.1 | 2 | 1640 | ||
NM_001318373.1 | NP_001305302.1 | 2 | 1640 | |||
NM_001318374.1 | NP_001305303.1 | 2 | 1640 | |||
NM_020403.4 | NP_065136.1 | 2 | 1640 | |||
NM_203487.2 | NP_982354.1 | 2 | 1640 | |||
XM_005266408.3 | XP_005266465.1 | 2 | 1640 | |||
XM_011535099.2 | XP_011533401.1 | 2 | 1640 | |||
XM_017020619.1 | XP_016876108.1 | 2 | 1408 | |||
XM_017020620.1 | XP_016876109.1 | 2 | 1640 | |||
XM_017020621.1 | XP_016876110.1 | 2 | 1407 | |||
GenBank mRNA | AF169692.2 | - | 1 | 1066 | ||
AK054661.1 | - | 2 | 1640 | |||
AK057768.1 | - | 1 | 550 | |||
AL834258.1 | - | 1 | 491 | |||
BC136626.1 | - | 2 | 1122 | 9020616 | ||
BC136627.1 | - | 2 | 1122 | 9020617 | ||
BC144321.1 | - | 2 | 1122 | 9052841 | ||
BC150296.1 | - | 2 | 1749 | 8860363 | ||
BX537422.1 | - | 2 | 1638 | |||
KJ534915.1 | - | 1 | 948 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
