siRNA Details
Gene Symbol: PCDH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318372.1 | NP_001305301.1 | 2 | 2449 | ||
NM_001318373.1 | NP_001305302.1 | 2 | 2449 | |||
NM_001318374.1 | NP_001305303.1 | 2 | 2449 | |||
NM_020403.4 | NP_065136.1 | 2 | 2449 | |||
NM_203487.2 | NP_982354.1 | 2 | 2449 | |||
XM_005266408.3 | XP_005266465.1 | 2 | 2449 | |||
XM_011535099.2 | XP_011533401.1 | 2 | 2449 | |||
XM_017020619.1 | XP_016876108.1 | 2 | 2217 | |||
XM_017020620.1 | XP_016876109.1 | 2 | 2449 | |||
XM_017020621.1 | XP_016876110.1 | 2 | 2216 | |||
GenBank mRNA | AF169692.2 | - | 1 | 1875 | ||
AK054661.1 | - | 2 | 2449 | |||
AK057768.1 | - | 1 | 1359 | |||
AL834258.1 | - | 1 | 1300 | |||
BC136626.1 | - | 2 | 1931 | 9020616 | ||
BC136627.1 | - | 2 | 1931 | 9020617 | ||
BC144321.1 | - | 2 | 1931 | 9052841 | ||
BC150296.1 | - | 2 | 2558 | 8860363 | ||
BX537422.1 | - | 2 | 2447 | |||
KJ534915.1 | - | 1 | 1757 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001271798, NM_001271800, NM_001271799, NM_001081377, AK147451, and others.
Gene Ontology Categories:
