siRNA Details
Gene Symbol: HIST4H4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175054.2 | NP_778224.1 | 1 | 313 | ||
GenBank mRNA | AK311916.1 | - | 1 | 313 | ||
BC020884.1 | - | 1 | 313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
