siRNA Details
Gene Symbol: SFMBT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016329.3 | NP_057413.2 | 5 | 806 | ||
XM_005265221.3 | XP_005265278.1 | 5 | 922 | |||
XM_006713203.3 | XP_006713266.1 | 6 | 1044 | |||
XM_006713204.3 | XP_006713267.1 | 5 | 744 | |||
XM_011533824.2 | XP_011532126.1 | 6 | 920 | |||
XM_011533825.2 | XP_011532127.1 | 6 | 767 | |||
GenBank mRNA | AB189472.1 | - | 4 | 430 | ||
AF168132.1 | - | 5 | 743 | |||
AK313965.1 | - | 5 | 801 | |||
AL080140.1 | - | 2 | 84 | |||
BC014614.2 | - | 5 | 690 | 3029598 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
