siRNA Details
Gene Symbol: SFMBT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016329.3 | NP_057413.2 | 4 | 671 | ||
XM_005265221.3 | XP_005265278.1 | 4 | 787 | |||
XM_006713203.3 | XP_006713266.1 | 5 | 909 | |||
XM_006713204.3 | XP_006713267.1 | 4 | 609 | |||
XM_011533824.2 | XP_011532126.1 | 5 | 785 | |||
XM_011533825.2 | XP_011532127.1 | 5 | 632 | |||
GenBank mRNA | AB189472.1 | - | 3 | 295 | ||
AF168132.1 | - | 4 | 608 | |||
AK313965.1 | - | 4 | 666 | |||
BC014614.2 | - | 4 | 555 | 3029598 | ||
BG723139.1 | - | 4 | 639 | |||
BU553665.1 | - | 5 | 774 | |||
DB081417.1 | - | 4 | 430 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
