siRNA Details
Gene Symbol: ZNF483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007169.3 | NP_001007170.1 | 3 | 666 | ||
NM_133464.3 | NP_597721.2 | 3 | 666 | |||
XM_011518300.2 | XP_011516602.1 | 3 | 1906 | |||
XM_017014337.1 | XP_016869826.1 | 4 | 879 | |||
XM_017014338.1 | XP_016869827.1 | 4 | 765 | |||
XM_017014339.1 | XP_016869828.1 | 3 | 666 | |||
GenBank mRNA | AB075842.1 | - | 3 | 592 | ||
AK092811.1 | - | 3 | 666 | |||
AK291267.1 | - | 3 | 658 | |||
BC065738.1 | - | 3 | 636 | |||
BC136695.1 | - | 2 | 459 | 9020685 | ||
BC136696.1 | - | 2 | 459 | 9020686 | ||
BC152427.1 | - | 3 | 592 | 8862541 | ||
BC152433.1 | - | 3 | 592 | 8862552 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
