siRNA Details
Gene Symbol: KIR3DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322168.1 | NP_001309097.1 | 4 | 616 | ||
NM_013289.2 | NP_037421.2 | 4 | 616 | |||
GenBank mRNA | AF022049.1 | - | 4 | 553 | ||
AF103010.1 | - | 4 | 553 | |||
AF103011.1 | - | 4 | 553 | |||
AF103012.1 | - | 4 | 553 | |||
AF103013.1 | - | 4 | 553 | |||
AF208688.1 | - | 2 | 490 | |||
AF208689.1 | - | 2 | 490 | |||
AF262968.1 | - | 2 | 472 | |||
AF262969.1 | - | 2 | 472 | |||
AF262970.1 | - | 2 | 472 | |||
AF262971.1 | - | 2 | 472 | |||
AF262973.1 | - | 4 | 586 | |||
AF262974.1 | - | 4 | 586 | |||
AJ417556.1 | - | 2 | 502 | |||
AJ417557.1 | - | 2 | 502 | |||
AY366236.1 | - | 4 | 591 | |||
AY366249.1 | - | 4 | 553 | |||
AY366250.1 | - | 4 | 579 | |||
AY366251.1 | - | 4 | 553 | |||
AY366252.1 | - | 4 | 553 | |||
AY760050.1 | - | 4 | 553 | |||
AY789065.1 | - | 4 | 568 | |||
AY789066.1 | - | 4 | 553 | |||
AY789072.1 | - | 4 | 553 | |||
BC028206.1 | - | 4 | 580 | 5215674 | ||
BC101540.1 | - | 4 | 572 | 8069046 | ||
BC101542.1 | - | 4 | 572 | 8069048 | ||
DQ178659.1 | - | 4 | 553 | |||
EF462885.1 | - | 2 | 483 | |||
EF472674.1 | - | 2 | 483 | |||
EF472675.1 | - | 2 | 483 | |||
EF472676.1 | - | 2 | 483 | |||
EF472677.1 | - | 2 | 483 | |||
EF472678.1 | - | 2 | 483 | |||
L41269.1 | - | 4 | 572 | |||
L76664.1 | - | 3 | 279 | |||
U30273.1 | - | 4 | 555 | |||
U30274.1 | - | 4 | 616 | |||
U31416.1 | - | 4 | 578 | |||
U33328.1 | - | 4 | 578 | |||
X94262.1 | - | 4 | 554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
