siRNA Details
Gene Symbol: KIAA1161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020702.4 | NP_065753.2 | 2 | 5511 | ||
XM_011517966.2 | XP_011516268.1 | 2 | 5562 | |||
XM_017014930.1 | XP_016870419.1 | 2 | 6111 | |||
GenBank mRNA | AB032987.2 | - | 1 | 5241 |
Gene Symbol: TSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015362.1 | 1 | 1082 | |||
GenBank mRNA | AY314745.1 | - | 1 | 1082 | ||
AY314746.1 | - | 1 | 1082 | |||
AY314747.1 | - | 1 | 1163 | |||
AY314748.1 | - | 1 | 1082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
