siRNA Details
Gene Symbol: HLA-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242758.1 | NP_001229687.1 | 2 | 178 | ||
NM_002116.7 | NP_002107.3 | 2 | 178 | |||
GenBank mRNA | AB209117.1 | - | 2 | 92 | ||
AF008933.1 | - | 2 | 88 | |||
AF012766.1 | - | 1 | 21 | |||
AF015930.1 | - | 2 | 94 | |||
AF036921.1 | - | 2 | 94 | |||
AF116215.1 | - | 2 | 94 | |||
AF140506.1 | - | 2 | 94 | |||
AF217561.1 | - | 2 | 94 | |||
AF287958.1 | - | 2 | 94 | |||
AF287959.1 | - | 2 | 94 | |||
AJ001269.1 | - | 1 | 21 | |||
AJ249241.1 | - | 2 | 94 | |||
AK130165.1 | - | 2 | 133 | |||
AK301014.1 | - | 2 | 120 | |||
AK301019.1 | - | 2 | 116 | |||
AK301282.1 | - | 2 | 116 | |||
AK304472.1 | - | 2 | 72 | |||
AK313119.1 | - | 2 | 116 | |||
AY191309.1 | - | 1 | 22 | |||
AY191310.1 | - | 1 | 22 | |||
AY365426.1 | - | 2 | 94 | |||
BC003069.2 | - | 2 | 95 | 3510139 | ||
BC019236.2 | - | 2 | 99 | 2821717 | ||
D38525.1 | - | 2 | 94 | |||
D83515.1 | - | 2 | 94 | |||
DQ336693.1 | - | 2 | 94 | |||
HG327114.1 | - | 2 | 94 | |||
KJ534864.1 | - | 2 | 94 | |||
L18898.1 | - | 2 | 94 | |||
L76936.1 | - | 1 | 21 | |||
M84377.1 | - | 2 | 94 | |||
M84378.1 | - | 2 | 94 | |||
M84379.1 | - | 2 | 94 | |||
M86404.1 | - | 2 | 70 | |||
U03754.1 | - | 2 | 94 | |||
U03907.1 | - | 2 | 94 | |||
U18930.1 | - | 2 | 94 | |||
U32184.1 | - | 2 | 94 | |||
X57954.1 | - | 2 | 70 | |||
X60764.1 | - | 2 | 70 | |||
X61700.1 | - | 2 | 94 | |||
X94566.1 | - | 1 | 17 | |||
X94567.1 | - | 1 | 17 | |||
X94568.1 | - | 1 | 17 | |||
X94573.1 | - | 1 | 17 | |||
X96724.1 | - | 2 | 94 | |||
Y11441.1 | - | 2 | 94 | |||
Y13028.1 | - | 1 | 21 | |||
Y13267.1 | - | 2 | 94 | |||
Z27120.1 | - | 2 | 94 | |||
Z46633.1 | - | 2 | 94 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
