siRNA Details
Gene Symbol: HLA-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242758.1 | NP_001229687.1 | 6 | 1108 | ||
NM_002116.7 | NP_002107.3 | 6 | 1108 | |||
GenBank mRNA | AB209117.1 | - | 6 | 1022 | ||
AF165065.1 | - | 6 | 1024 | |||
AF217561.1 | - | 6 | 1024 | |||
AK301019.1 | - | 6 | 1240 | |||
AK301282.1 | - | 7 | 970 | |||
AK304472.1 | - | 6 | 1002 | |||
AK313119.1 | - | 6 | 1046 | |||
AK316195.1 | - | 4 | 1663 | |||
AY786585.1 | - | 6 | 1024 | |||
AY786586.1 | - | 6 | 1042 | |||
AY786587.1 | - | 6 | 1042 | |||
AY786588.1 | - | 5 | 1265 | |||
BC003069.2 | - | 6 | 1025 | 3510139 | ||
BE875389.1 | - | 1 | 22 | |||
D16841.1 | - | 6 | 1024 | |||
D16842.1 | - | 6 | 1024 | |||
DQ327721.1 | - | 6 | 1024 | |||
DQ336694.1 | - | 6 | 1042 | |||
DQ336695.1 | - | 6 | 1024 | |||
HG327114.1 | - | 6 | 1024 | |||
M27971.1 | - | 3 | 280 | |||
U07161.1 | - | 6 | 1024 | |||
U32184.1 | - | 6 | 1024 | |||
U50574.1 | - | 6 | 1024 | |||
X13111.1 | - | 6 | 1024 | |||
X61700.1 | - | 6 | 1024 | |||
Y17224.1 | - | 6 | 1042 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
