siRNA Details
Gene Symbol: HLA-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002116.7 | NP_002107.3 | 2 | 347 | ||
GenBank mRNA | AF015930.1 | - | 2 | 263 | ||
AF165065.1 | - | 2 | 263 | |||
AF217561.1 | - | 2 | 263 | |||
AF287958.1 | - | 2 | 263 | |||
AF287959.1 | - | 2 | 263 | |||
AK301014.1 | - | 2 | 289 | |||
AK301019.1 | - | 2 | 285 | |||
AK301282.1 | - | 2 | 285 | |||
AK304472.1 | - | 2 | 241 | |||
AK313119.1 | - | 2 | 285 | |||
AK316195.1 | - | 2 | 219 | |||
AY786585.1 | - | 2 | 263 | |||
AY786586.1 | - | 2 | 263 | |||
AY786587.1 | - | 2 | 263 | |||
AY786588.1 | - | 2 | 263 | |||
D16841.1 | - | 2 | 263 | |||
D16842.1 | - | 2 | 263 | |||
DQ327718.1 | - | 2 | 263 | |||
DQ327721.1 | - | 2 | 263 | |||
DQ336694.1 | - | 2 | 263 | |||
DQ336695.1 | - | 2 | 263 | |||
HG327114.1 | - | 2 | 263 | |||
KJ534864.1 | - | 2 | 263 | |||
M84375.1 | - | 2 | 263 | |||
U03907.1 | - | 2 | 263 | |||
U32184.1 | - | 2 | 263 | |||
U50574.1 | - | 2 | 263 | |||
X13111.1 | - | 2 | 263 | |||
X13112.1 | - | 2 | 263 | |||
X91399.1 | - | 1 | 190 | |||
Y08275.1 | - | 1 | 190 | |||
Y17224.1 | - | 2 | 263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
