siRNA Details
Gene Symbol: ICA1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288622.1 | NP_001275551.1 | 2 | 407 | ||
NM_001288623.1 | NP_001275552.1 | 2 | 352 | |||
NM_001288624.1 | NP_001275553.1 | 1 | 258 | |||
NM_138468.5 | NP_612477.3 | 3 | 534 | |||
GenBank mRNA | AB053316.1 | - | 4 | 410 | ||
AK055978.1 | - | 2 | 149 | |||
AK092310.1 | - | 2 | 407 | |||
AL833904.1 | - | 3 | 191 | |||
BC000993.2 | - | 1 | 258 | |||
BC063876.1 | - | 2 | 65 | 30377894 | ||
BC093902.1 | - | 2 | 69 | |||
BC112235.1 | - | 2 | 69 | |||
DB097704.1 | - | 4 | 270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
