siRNA Details
Gene Symbol: SEC1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004401.2 | 5 | 562 | |||
GenBank mRNA | AK301046.1 | - | 3 | 251 | ||
AK301238.1 | - | 5 | 502 | |||
D87946.1 | - | 2 | 210 | |||
D87947.1 | - | 4 | 408 | |||
DC353776.1 | - | 3 | 313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
