siRNA Details
Gene Symbol: OSCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282349.1 | NP_001269278.1 | 4 | 343 | ||
NM_001282350.1 | NP_001269279.1 | 3 | 310 | |||
NM_130771.4 | NP_570127.3 | 5 | 355 | |||
NM_133168.4 | NP_573398.2 | 3 | 310 | |||
NM_133169.4 | NP_573399.2 | 4 | 343 | |||
NM_206818.2 | NP_996554.2 | 5 | 355 | |||
GenBank mRNA | AF251702.1 | - | 3 | 197 | ||
AF391162.1 | - | 3 | 158 | |||
AF391163.1 | - | 4 | 163 | |||
AF391164.1 | - | 2 | 156 | |||
AF474152.1 | - | 3 | 189 | |||
AF474153.1 | - | 2 | 118 | |||
AK091111.1 | - | 3 | 160 | |||
AK130199.1 | - | 4 | 187 | |||
AK311307.1 | - | 3 | 283 | |||
BC035023.2 | - | 4 | 344 | 4825982 | ||
BG722509.1 | - | 4 | 349 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
