siRNA Details
Gene Symbol: COL11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190709.1 | NP_001177638.1 | 26 | 2466 | ||
NM_001854.3 | NP_001845.3 | 27 | 2583 | |||
NM_080629.2 | NP_542196.2 | 27 | 2619 | |||
NM_080630.3 | NP_542197.3 | 25 | 2235 | |||
NR_134980.1 | 27 | 2583 | ||||
XM_017000334.1 | XP_016855823.1 | 28 | 2819 | |||
XM_017000335.1 | XP_016855824.1 | 28 | 2813 | |||
XM_017000336.1 | XP_016855825.1 | 28 | 2820 | |||
XM_017000337.1 | XP_016855826.1 | 21 | 1297 | |||
GenBank mRNA | AB208844.1 | - | 23 | 1583 | ||
BC117697.1 | - | 25 | 2218 | 40073305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
