siRNA Details
Gene Symbol: CBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007276.4 | NP_009207.2 | 6 | 1749 | ||
NM_016587.3 | NP_057671.2 | 6 | 1521 | |||
XM_005249611.3 | XP_005249668.1 | 6 | 2256 | |||
GenBank mRNA | AB030905.1 | - | 4 | 1336 | ||
AF136630.1 | - | 6 | 1442 | |||
BC000954.1 | - | 6 | 1477 | 3450099 | ||
BC014380.1 | - | 2 | 1037 | |||
BX647444.1 | - | 6 | 1382 | |||
BX648807.1 | - | 6 | 1477 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
