siRNA Details
Gene Symbol: PRR31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005266138.4 | XP_005266195.1 | 3 | 832 | ||
XM_011519282.2 | XP_011517584.1 | 4 | 920 | |||
GenBank mRNA | BI832003.1 | - | 3 | 298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
