siRNA Details
Gene Symbol: PRKAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276289.1 | NP_001263218.1 | 12 | 3619 | ||
NM_001278433.1 | NP_001265362.1 | 11 | 3489 | |||
NM_002734.4 | NP_002725.1 | 11 | 3486 | |||
NM_212471.2 | NP_997636.1 | 11 | 3679 | |||
NM_212472.2 | NP_997637.1 | 11 | 3517 | |||
XM_011524983.2 | XP_011523285.1 | 11 | 3582 | |||
XM_011524984.2 | XP_011523286.1 | 12 | 3520 | |||
XM_011524985.2 | XP_011523287.1 | 12 | 3814 | |||
GenBank mRNA | AK124586.1 | - | 11 | 3408 | ||
AL050038.1 | - | 1 | 851 | |||
AY007115.1 | - | 1 | 666 | |||
BC036285.1 | - | 11 | 3438 | 4340015 | ||
BC093042.1 | - | 11 | 3426 | 6514677 | ||
CR749311.1 | - | 12 | 3548 | |||
S54711.1 | - | 1 | 75 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
