siRNA Details
Gene Symbol: PRKAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276289.1 | NP_001263218.1 | 12 | 1760 | ||
NM_001278433.1 | NP_001265362.1 | 11 | 1630 | |||
NM_002734.4 | NP_002725.1 | 11 | 1627 | |||
NM_212471.2 | NP_997636.1 | 11 | 1820 | |||
NM_212472.2 | NP_997637.1 | 11 | 1658 | |||
XM_011524983.2 | XP_011523285.1 | 11 | 1723 | |||
XM_011524984.2 | XP_011523286.1 | 12 | 1661 | |||
XM_011524985.2 | XP_011523287.1 | 12 | 1955 | |||
GenBank mRNA | AK124586.1 | - | 11 | 1546 | ||
BC036285.1 | - | 11 | 1579 | 4340015 | ||
BC093042.1 | - | 11 | 1568 | 6514677 | ||
CR749311.1 | - | 12 | 1689 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
