siRNA Details
Gene Symbol: PRKAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276289.1 | NP_001263218.1 | 8 | 994 | ||
NM_001276290.1 | NP_001263219.1 | 6 | 691 | |||
NM_001278433.1 | NP_001265362.1 | 7 | 864 | |||
NM_002734.4 | NP_002725.1 | 7 | 861 | |||
NM_212471.2 | NP_997636.1 | 7 | 1054 | |||
NM_212472.2 | NP_997637.1 | 7 | 892 | |||
XM_011524983.2 | XP_011523285.1 | 7 | 957 | |||
XM_011524984.2 | XP_011523286.1 | 8 | 895 | |||
XM_011524985.2 | XP_011523287.1 | 8 | 1189 | |||
GenBank mRNA | A12295.1 | - | 7 | 787 | ||
AK097580.1 | - | 6 | 1264 | |||
AK124586.1 | - | 7 | 780 | |||
AK312307.1 | - | 7 | 790 | |||
AL535889.3 | - | 7 | 832 | |||
BC036285.1 | - | 7 | 813 | 4340015 | ||
BC093042.1 | - | 7 | 802 | 6514677 | ||
CR749311.1 | - | 8 | 923 | |||
M18468.1 | - | 7 | 788 | |||
M33336.1 | - | 7 | 772 | |||
S54707.1 | - | 3 | 217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
