siRNA Details
Gene Symbol: GYPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198682.2 | NP_941391.2 | 4 | 1243 | ||
XM_017008139.1 | XP_016863628.1 | 5 | 5170 | |||
GenBank mRNA | BC017864.1 | - | 4 | 1239 | 4686481 | |
BC064396.1 | - | 4 | 1200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
