siRNA Details
Gene Symbol: NUMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005743.1 | NP_001005743.1 | 4 | 382 | ||
NM_001005744.1 | NP_001005744.1 | 4 | 382 | |||
NM_001005745.1 | NP_001005745.1 | 4 | 382 | |||
NM_001320114.1 | NP_001307043.1 | 4 | 402 | |||
NM_003744.5 | NP_003735.3 | 4 | 382 | |||
GenBank mRNA | AF015040.1 | - | 4 | 340 | ||
AF108092.1 | - | 2 | 167 | |||
AF171938.1 | - | 4 | 332 | |||
AF171939.1 | - | 4 | 332 | |||
AF171940.1 | - | 4 | 332 | |||
AF171941.1 | - | 4 | 332 | |||
AK304193.1 | - | 2 | 185 | |||
AK315130.1 | - | 3 | 263 | |||
BC020788.1 | - | 3 | 263 | |||
BC068476.1 | - | 4 | 382 | 4822300 | ||
BX248073.1 | - | 3 | 239 | |||
BX248775.1 | - | 4 | 311 | |||
BX248781.1 | - | 3 | 277 | |||
BX647137.1 | - | 4 | 341 | |||
EU265734.1 | - | 2 | 101 | |||
EU265735.1 | - | 2 | 101 | |||
EU265736.1 | - | 2 | 101 | |||
EU265737.1 | - | 2 | 101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
