siRNA Details
Gene Symbol: SPACA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316972.1 | NP_001303901.1 | 8 | 998 | ||
NM_001316994.1 | NP_001303923.1 | 8 | 750 | |||
NR_024330.1 | 7 | 1737 | ||||
XM_017026299.1 | XP_016881788.1 | 8 | 2521 | |||
XM_017026300.1 | XP_016881789.1 | 8 | 881 | |||
XM_017026301.1 | XP_016881790.1 | 8 | 947 | |||
GenBank mRNA | AK024362.1 | - | 579 | |||
KF751883.1 | - | 8 | 839 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
