siRNA Details
Gene Symbol: OR5AC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054106.1 | NP_473447.1 | 1 | 787 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_146779.
Gene Ontology Categories:
