siRNA Details
Gene Symbol: ZBTB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278196.1 | NP_001265125.1 | 2 | 3152 | ||
NM_006352.4 | NP_006343.2 | 1 | 3680 | |||
NM_205768.2 | NP_991331.1 | 2 | 3283 | |||
XM_005273006.2 | XP_005273063.1 | 3 | 3213 | |||
XM_017000060.1 | XP_016855549.1 | 3 | 5313 | |||
GenBank mRNA | AJ001388.1 | - | 1 | 3607 | ||
AL050055.1 | - | 1 | 897 | |||
BC036677.2 | - | 2 | 3254 | 5259205 | ||
BC078175.1 | - | 1 | 3679 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
