siRNA Details
Gene Symbol: RFX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282116.1 | NP_001269045.1 | 14 | 2018 | ||
NM_002919.3 | NP_002910.1 | 13 | 1763 | |||
NM_134428.2 | NP_602304.1 | 15 | 2085 | |||
XM_006716840.1 | XP_006716903.1 | 14 | 1931 | |||
XM_006716843.1 | XP_006716906.1 | 15 | 1931 | |||
XM_006716844.1 | XP_006716907.1 | 15 | 1947 | |||
XM_006716845.2 | XP_006716908.1 | 14 | 2009 | |||
XM_006716847.3 | XP_006716910.1 | 9 | 1064 | |||
XM_011517998.1 | XP_011516300.1 | 15 | 1999 | |||
XM_011517999.1 | XP_011516301.1 | 15 | 1963 | |||
XM_011518000.1 | XP_011516302.1 | 13 | 1934 | |||
XM_011518001.1 | XP_011516303.1 | 16 | 2013 | |||
XM_017015000.1 | XP_016870489.1 | 15 | 1950 | |||
XM_017015001.1 | XP_016870490.1 | 12 | 1688 | |||
XM_017015002.1 | XP_016870491.1 | 8 | 3470 | |||
GenBank mRNA | AK289540.1 | - | 14 | 1816 | ||
BC022191.2 | - | 15 | 2066 | 3531443 | ||
X76092.1 | - | 13 | 1763 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
