siRNA Details
Gene Symbol: ZBTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123329.1 | NP_001116801.1 | 2 | 2346 | ||
XM_005267410.1 | XP_005267467.1 | 4 | 2482 | |||
XM_011536566.2 | XP_011534868.1 | 3 | 2219 | |||
XM_017021094.1 | XP_016876583.1 | 4 | 3358 | |||
XM_017021095.1 | XP_016876584.1 | 3 | 2274 | |||
GenBank mRNA | AK291743.1 | - | 2 | 2124 | ||
BC050719.1 | - | 4 | 2386 | 6141266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
