siRNA Details
Gene Symbol: SYN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135774.1 | NP_001129246.1 | 2 | 326 | ||
NM_003490.3 | NP_003481.3 | 1 | 223 | |||
NM_133633.2 | NP_598344.2 | 1 | 223 | |||
XM_011530405.2 | XP_011528707.1 | 2 | 859 | |||
XM_011530406.2 | XP_011528708.1 | 2 | 325 | |||
XM_011530407.2 | XP_011528709.1 | 2 | 732 | |||
XM_011530408.1 | XP_011528710.1 | 2 | 323 | |||
XM_017028960.1 | XP_016884449.1 | 3 | 1257 | |||
XM_017028961.1 | XP_016884450.1 | 2 | 649 | |||
XM_017028962.1 | XP_016884451.1 | 2 | 584 | |||
XM_017028963.1 | XP_016884452.1 | 2 | 248 | |||
XM_017028964.1 | XP_016884453.1 | 2 | 323 | |||
GenBank mRNA | AB209684.1 | - | 2 | 278 | ||
AF046873.1 | - | 1 | 100 | |||
BC075065.2 | - | 1 | 90 | 30915588 | ||
BC075066.2 | - | 1 | 90 | 30915375 | ||
BC117460.1 | - | 1 | 91 | 40126011 | ||
BC143874.1 | - | 1 | 91 | 9052390 | ||
CR456589.1 | - | 1 | 155 | |||
DB458758.1 | - | 2 | 323 | |||
DQ655947.2 | - | 2 | 550 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
