siRNA Details
Gene Symbol: SYN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135774.1 | NP_001129246.1 | 6 | 902 | ||
NM_003490.3 | NP_003481.3 | 5 | 802 | |||
NM_133633.2 | NP_598344.2 | 5 | 802 | |||
XM_011530405.2 | XP_011528707.1 | 6 | 1438 | |||
XM_011530406.2 | XP_011528708.1 | 6 | 904 | |||
XM_011530407.2 | XP_011528709.1 | 6 | 1311 | |||
XM_011530408.1 | XP_011528710.1 | 6 | 902 | |||
XM_011530410.2 | XP_011528712.1 | 4 | 1106 | |||
XM_017028960.1 | XP_016884449.1 | 7 | 1836 | |||
XM_017028961.1 | XP_016884450.1 | 6 | 1228 | |||
XM_017028962.1 | XP_016884451.1 | 6 | 1163 | |||
XM_017028963.1 | XP_016884452.1 | 6 | 827 | |||
XM_017028964.1 | XP_016884453.1 | 6 | 899 | |||
GenBank mRNA | AB209684.1 | - | 6 | 854 | ||
AF046873.1 | - | 5 | 679 | |||
BC075065.2 | - | 5 | 669 | 30915588 | ||
BC075066.2 | - | 5 | 669 | 30915375 | ||
BC117460.1 | - | 5 | 667 | 40126011 | ||
BC143874.1 | - | 5 | 670 | 9052390 | ||
CR456589.1 | - | 5 | 734 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
