siRNA Details
Gene Symbol: THAP7-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027051.1 | 1 | 653 | |||
NR_027052.1 | 1 | 653 | ||||
GenBank mRNA | AK096901.1 | - | 1 | 358 | ||
BC110368.1 | - | 1 | 666 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
