siRNA Details
Gene Symbol: ZKSCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287054.1 | NP_001273983.1 | 8 | 3784 | ||
NM_001287055.1 | NP_001273984.1 | 4 | 3087 | |||
NM_003439.2 | NP_003430.1 | 6 | 3755 | |||
GenBank mRNA | AK074116.1 | - | 6 | 3713 | ||
AK091471.1 | - | 4 | 3056 | |||
AK123012.1 | - | 1 | 3757 | |||
AL833578.1 | - | 6 | 3731 | |||
BC011421.1 | - | 1 | 1760 | |||
BX640646.1 | - | 6 | 3728 | |||
U09848.1 | - | 2 | 2797 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
