siRNA Details
Gene Symbol: HIST1H4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003538.3 | NP_003529.1 | 1 | 18 | ||
GenBank mRNA | BC066248.1 | - | 1 | 18 | 7002088 | |
BC066249.1 | - | 1 | 18 | 7002089 | ||
BC067495.1 | - | 1 | 18 | 7002090 | ||
BC069654.1 | - | 1 | 18 | 7262752 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
