siRNA Details
Gene Symbol: TNRC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014494.2 | NP_055309.2 | 17 | 4488 | ||
XM_005255254.3 | XP_005255311.1 | 16 | 4331 | |||
XM_005255257.4 | XP_005255314.1 | 16 | 4455 | |||
XM_017023143.1 | XP_016878632.1 | 16 | 4432 | |||
XM_017023144.1 | XP_016878633.1 | 15 | 4399 | |||
XM_017023145.1 | XP_016878634.1 | 16 | 4365 | |||
XM_017023146.1 | XP_016878635.1 | 16 | 4497 | |||
XM_017023147.1 | XP_016878636.1 | 16 | 4525 | |||
XM_017023148.1 | XP_016878637.1 | 15 | 4252 | |||
XM_017023149.1 | XP_016878638.1 | 14 | 4249 | |||
XM_017023150.1 | XP_016878639.1 | 15 | 4214 | |||
XM_017023151.1 | XP_016878640.1 | 14 | 4103 | |||
XM_017023152.1 | XP_016878641.1 | 15 | 4004 | |||
XM_017023153.1 | XP_016878642.1 | 16 | 4455 | |||
XM_017023154.1 | XP_016878643.1 | 14 | 4346 | |||
GenBank mRNA | AB040893.1 | - | 12 | 2688 | ||
AJ492221.1 | - | 16 | 4374 | |||
AK025696.1 | - | 10 | 1045 | |||
AK094542.1 | - | 4 | 1070 | |||
AL713750.1 | - | 4 | 1241 | |||
AL833757.1 | - | 11 | 1649 | |||
AY035864.1 | - | 14 | 4022 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
