siRNA Details
Gene Symbol: TNRC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014494.2 | NP_055309.2 | 17, 18 | 4660 | ||
XM_005255254.3 | XP_005255311.1 | 16, 17 | 4503 | |||
XM_005255257.4 | XP_005255314.1 | 16, 17 | 4627 | |||
XM_017023143.1 | XP_016878632.1 | 16, 17 | 4604 | |||
XM_017023144.1 | XP_016878633.1 | 15, 16 | 4571 | |||
XM_017023145.1 | XP_016878634.1 | 16, 17 | 4537 | |||
XM_017023146.1 | XP_016878635.1 | 16, 17 | 4669 | |||
XM_017023147.1 | XP_016878636.1 | 16, 17 | 4697 | |||
XM_017023148.1 | XP_016878637.1 | 15, 16 | 4424 | |||
XM_017023149.1 | XP_016878638.1 | 14, 15 | 4421 | |||
XM_017023150.1 | XP_016878639.1 | 15, 16 | 4386 | |||
XM_017023151.1 | XP_016878640.1 | 14, 15 | 4275 | |||
XM_017023152.1 | XP_016878641.1 | 15, 16 | 4176 | |||
XM_017023153.1 | XP_016878642.1 | 16, 17 | 4627 | |||
XM_017023154.1 | XP_016878643.1 | 14, 15 | 4518 | |||
GenBank mRNA | AB040893.1 | - | 12, 13 | 2860 | ||
AJ492221.1 | - | 16, 17 | 4546 | |||
AK025696.1 | - | 10, 11 | 1217 | |||
AK094542.1 | - | 4, 5 | 1242 | |||
AL713750.1 | - | 4, 5 | 1413 | |||
AL833757.1 | - | 11, 12 | 1821 | |||
AY035864.1 | - | 14, 15 | 4194 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_144925, AK142809, AK142808, BC005741, AK044679, and others.
Gene Ontology Categories:
