siRNA Details
Gene Symbol: TNRC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014494.2 | NP_055309.2 | 18, 19 | 4786 | ||
XM_005255254.3 | XP_005255311.1 | 17, 18 | 4629 | |||
XM_005255257.4 | XP_005255314.1 | 17, 18 | 4753 | |||
XM_017023143.1 | XP_016878632.1 | 17, 18 | 4730 | |||
XM_017023144.1 | XP_016878633.1 | 16, 17 | 4697 | |||
XM_017023145.1 | XP_016878634.1 | 17, 18 | 4663 | |||
XM_017023146.1 | XP_016878635.1 | 17, 18 | 4795 | |||
XM_017023147.1 | XP_016878636.1 | 17, 18 | 4823 | |||
XM_017023148.1 | XP_016878637.1 | 16, 17 | 4550 | |||
XM_017023149.1 | XP_016878638.1 | 15, 16 | 4547 | |||
XM_017023150.1 | XP_016878639.1 | 16, 17 | 4512 | |||
XM_017023151.1 | XP_016878640.1 | 15, 16 | 4401 | |||
XM_017023152.1 | XP_016878641.1 | 16, 17 | 4302 | |||
XM_017023153.1 | XP_016878642.1 | 17, 18 | 4753 | |||
XM_017023154.1 | XP_016878643.1 | 15, 16 | 4644 | |||
GenBank mRNA | AB040893.1 | - | 13, 14 | 2986 | ||
AJ492221.1 | - | 17, 18 | 4672 | |||
AK001773.1 | - | 1, 2 | 111 | |||
AK025696.1 | - | 11, 12 | 1343 | |||
AK094542.1 | - | 5, 6 | 1368 | |||
AL713750.1 | - | 5, 6 | 1539 | |||
AL833757.1 | - | 12, 13 | 1947 | |||
AY035864.1 | - | 15, 16 | 4320 | |||
BC024324.1 | - | 1, 2 | 132 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_144925, AK142809, AK142808, BC005741, AK044679, and others.
Gene Ontology Categories:
