siRNA Details
Gene Symbol: AP2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014203.2 | NP_055018.2 | 3, 4 | 499 | ||
NM_130787.2 | NP_570603.2 | 3, 4 | 499 | |||
XM_011526556.2 | XP_011524858.1 | 3, 4 | 1109 | |||
XM_011526557.2 | XP_011524859.1 | 3, 4 | 1104 | |||
GenBank mRNA | AK124343.1 | - | 3, 4 | 415 | ||
AK302005.1 | - | 3, 4 | 454 | |||
AL136925.1 | - | 3, 4 | 365 | |||
BC014214.2 | - | 3, 4 | 397 | 4111165 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
