siRNA Details
Gene Symbol: KANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136191.2 | NP_001129663.1 | 13 | 3977 | ||
NM_015493.6 | NP_056308.3 | 11 | 3719 | |||
GenBank mRNA | AB040951.2 | - | 12 | 3929 | ||
AB284125.1 | - | 14 | 4160 | |||
AL117489.1 | - | 1 | 733 | |||
CR457388.1 | - | 1 | 40 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
