siRNA Details
Gene Symbol: LGALS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190441.2 | NP_001177370.2 | 3 | 247 | ||
GenBank mRNA | BX095861.1 | - | 3 | 207 | ||
FJ613352.1 | - | 3 | 247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
