siRNA Details
Gene Symbol: DBF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025104.3 | NP_079380.2 | 13 | 1310 | ||
NM_145663.2 | NP_663696.1 | 13 | 1310 | |||
NR_036623.1 | 12 | 1222 | ||||
XM_006722098.2 | XP_006722161.1 | 13 | 1283 | |||
XM_006722102.3 | XP_006722165.1 | 11 | 1185 | |||
XM_011525281.2 | XP_011523583.1 | 12 | 2424 | |||
XM_017025145.1 | XP_016880634.1 | 13 | 1424 | |||
XM_017025146.1 | XP_016880635.1 | 13 | 1316 | |||
XM_017025147.1 | XP_016880636.1 | 12 | 1415 | |||
XM_017025148.1 | XP_016880637.1 | 12 | 1307 | |||
XM_017025149.1 | XP_016880638.1 | 13 | 1424 | |||
XM_017025150.1 | XP_016880639.1 | 12 | 1166 | |||
XM_017025152.1 | XP_016880641.1 | 10 | 4047 | |||
XM_017025153.1 | XP_016880642.1 | 11 | 2218 | |||
XM_017025154.1 | XP_016880643.1 | 10 | 880 | |||
XM_017025155.1 | XP_016880644.1 | 11 | 1120 | |||
GenBank mRNA | AF448801.1 | - | 13 | 1235 | ||
AF465820.1 | - | 7 | 972 | |||
AK125771.1 | - | 4 | 340 | |||
BC016158.1 | - | 13 | 1310 | 3536735 | ||
BC033660.2 | - | 12 | 1144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
