siRNA Details
Gene Symbol: DBF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025104.3 | NP_079380.2 | 11, 12 | 1151 | ||
NM_145663.2 | NP_663696.1 | 11, 12 | 1151 | |||
NR_036623.1 | 10, 11 | 1063 | ||||
XM_006722098.2 | XP_006722161.1 | 11, 12 | 1124 | |||
XM_006722102.3 | XP_006722165.1 | 9, 10 | 1026 | |||
XM_011525281.2 | XP_011523583.1 | 10, 11 | 2265 | |||
XM_017025145.1 | XP_016880634.1 | 11, 12 | 1265 | |||
XM_017025146.1 | XP_016880635.1 | 11, 12 | 1157 | |||
XM_017025147.1 | XP_016880636.1 | 10, 11 | 1256 | |||
XM_017025148.1 | XP_016880637.1 | 10, 11 | 1148 | |||
XM_017025149.1 | XP_016880638.1 | 11, 12 | 1265 | |||
XM_017025150.1 | XP_016880639.1 | 10, 11 | 1007 | |||
XM_017025151.1 | XP_016880640.1 | 11, 12 | 1265 | |||
XM_017025152.1 | XP_016880641.1 | 8, 9 | 3888 | |||
XM_017025153.1 | XP_016880642.1 | 9, 10 | 2059 | |||
XM_017025154.1 | XP_016880643.1 | 8, 9 | 721 | |||
XM_017025155.1 | XP_016880644.1 | 9, 10 | 961 | |||
GenBank mRNA | AF448801.1 | - | 11, 12 | 1076 | ||
AF465820.1 | - | 5, 6 | 813 | |||
AK125771.1 | - | 2, 3 | 181 | |||
BC016158.1 | - | 11, 12 | 1151 | 3536735 | ||
BC033660.2 | - | 10, 11 | 985 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
